siRNA Details
| Gene Symbol: Zcchc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138590.4 | NP_613056.2 | 8 | 1316 | ||
| GenBank mRNA | AK013175.1 | - | 2 | 800 | ||
| AK050430.1 | - | 2 | 2008 | |||
| BC050200.1 | - | 7 | 1191 | |||
| BC054405.1 | - | 2 | 519 | |||

Zcchc7
Genomic Map