siRNA Details
| Gene Symbol: Usp39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138592.4 | NP_613058.1 | 10 | 1435 | ||
| GenBank mRNA | AK010443.1 | - | 10 | 1445 | ||
| AK018595.1 | - | 10 | 1436 | |||
| AK033105.1 | - | 10 | 1432 | |||
| AK045062.1 | - | 10 | 1432 | |||
| AK088354.1 | - | 10 | 1447 | |||
| AK165625.1 | - | 10 | 1416 | |||
| AK167668.1 | - | 10 | 1423 | |||
| BC026983.1 | - | 10 | 1410 | 5365862 | ||

Usp39
Genomic Map