siRNA Details
Gene Symbol: D6Wsu163e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138594.3 | NP_613060.1 | 11 | 1416 | ||
GenBank mRNA | AK019434.1 | - | 3 | 306 | ||
AK037095.1 | - | 11 | 1421 | |||
AK045541.1 | - | 11 | 1417 | |||
AK046707.1 | - | 11 | 1404 | |||
AK137201.1 | - | 11 | 1407 | |||
BC016429.1 | - | 10 | 1364 | 4486831 |
