siRNA Details
Gene Symbol: D6Wsu163e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138594.3 | NP_613060.1 | 13, 14 | 1636 | ||
GenBank mRNA | AK019434.1 | - | 5, 6 | 526 | ||
AK045541.1 | - | 13, 14 | 1637 | |||
AK046707.1 | - | 13, 14 | 1624 | |||
AK137201.1 | - | 13, 14 | 1627 | |||
BC016429.1 | - | 12, 13 | 1584 | 4486831 |
