siRNA Details
| Gene Symbol: Emilin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182840.2 | NP_878260.1 | 4 | 724 | ||
| GenBank mRNA | BC054738.1 | - | 4 | 713 | 6402350 | |
| BC057619.1 | - | 4 | 713 | 6402350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emilin3| Gene Symbol: Emilin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182840.2 | NP_878260.1 | 4 | 724 | ||
| GenBank mRNA | BC054738.1 | - | 4 | 713 | 6402350 | |
| BC057619.1 | - | 4 | 713 | 6402350 | ||