siRNA Details
| Gene Symbol: B3gat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172124.2 | NP_742122.2 | 1 | 754 | ||
| GenBank mRNA | AB055902.1 | - | 1 | 700 | ||
| AK048146.1 | - | 1 | 670 | |||
| AK052640.1 | - | 1 | 754 | |||
| AK220433.1 | - | 662 | ||||
| BC056368.1 | - | 1 | 646 | 6827380 | ||
| BC058082.1 | - | 1 | 646 | 6827380 | ||

B3gat2
Genomic Map