siRNA Details
Gene Symbol: Rtcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145422.4 | NP_663397.1 | 7 | 823 | ||
GenBank mRNA | AK145891.1 | - | 7 | 820 | ||
AK145916.1 | - | 7 | 820 | |||
AK149885.1 | - | 7 | 778 | |||
AK150376.1 | - | 7 | 782 | |||
AK150470.1 | - | 7 | 782 | |||
AK151042.1 | - | 7 | 816 | |||
AK151182.1 | - | 7 | 782 | |||
AK151932.1 | - | 7 | 782 | |||
AK151988.1 | - | 7 | 816 | |||
AK152144.1 | - | 7 | 782 | |||
AK153434.1 | - | 7 | 782 | |||
AK167643.1 | - | 7 | 809 | |||
AK168038.1 | - | 7 | 817 | |||
AK172175.1 | - | 7 | 817 | |||
AY424364.1 | - | 7 | 816 | |||
BC003288.1 | - | 7 | 825 | 3583081 |
