siRNA Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170855.1 | NP_001164326.1 | 9 | 1862 | ||
| NM_178872.4 | NP_849203.2 | 9 | 1863 | |||
| GenBank mRNA | AB103063.1 | - | 9 | 1657 | ||
| AY251389.1 | - | 9 | 1620 | |||
| BC058106.1 | - | 9 | 1781 | 6836197 | ||

Trim36
Genomic Map