siRNA Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170855.1 | NP_001164326.1 | 8 | 1550 | ||
| NM_178872.4 | NP_849203.2 | 8 | 1551 | |||
| GenBank mRNA | AB103063.1 | - | 8 | 1345 | ||
| AY251389.1 | - | 8 | 1308 | |||
| BC058106.1 | - | 8 | 1469 | 6836197 | ||

Trim36
Genomic Map