siRNA Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145705.3 | NP_663751.2 | 8 | 1341 | ||
| GenBank mRNA | AF518764.1 | - | 8 | 1306 | ||
| AI844697.1 | - | 2 | 395 | |||
| AK011154.1 | - | 8 | 1309 | |||
| AK016456.1 | - | 8 | 1275 | |||
| AK028952.1 | - | 7 | 1466 | |||
| AK090324.1 | - | 8 | 1308 | |||
| AK144592.1 | - | 8 | 1270 | |||
| AK169480.1 | - | 3 | 1355 | |||
| AK214714.1 | - | 2 | 48 | |||
| BC017142.1 | - | 3 | 1757 | |||
| BC030347.1 | - | 8 | 1267 | 5003804 | ||

Tinf2
Genomic Map