siRNA Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204915.1 | NP_001191844.1 | 7 | 796 | ||
| NM_178606.5 | NP_848721.1 | 7 | 796 | |||
| GenBank mRNA | AK005026.1 | - | 7 | 742 | ||
| AK012403.1 | - | 7 | 740 | |||
| AK028868.1 | - | 7 | 753 | |||
| AK047728.1 | - | 7 | 664 | |||
| AK079649.1 | - | 7 | 666 | |||
| AK154559.1 | - | 7 | 654 | |||
| AY562231.1 | - | 7 | 629 | |||
| BC004607.1 | - | 7 | 705 | 3585133 | ||

Reep3
Genomic Map