siRNA Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204915.1 | NP_001191844.1 | 5 | 505 | ||
| NM_178606.5 | NP_848721.1 | 5 | 505 | |||
| GenBank mRNA | AK012403.1 | - | 5 | 449 | ||
| AK028868.1 | - | 5 | 462 | |||
| AK047728.1 | - | 5 | 373 | |||
| AK079649.1 | - | 5 | 375 | |||
| AK154559.1 | - | 5 | 363 | |||
| AY562231.1 | - | 5 | 338 | |||
| BC004607.1 | - | 5 | 414 | 3585133 | ||

Reep3
Genomic Map