siRNA Details
| Gene Symbol: Apon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133996.3 | NP_598757.1 | 2 | 729 | ||
| GenBank mRNA | AK083289.1 | - | 1 | 865 | ||
| BC022137.1 | - | 1 | 796 | |||
| BC030734.1 | - | 1 | 669 | |||
| BC034832.1 | - | 1 | 828 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apon| Gene Symbol: Apon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133996.3 | NP_598757.1 | 2 | 729 | ||
| GenBank mRNA | AK083289.1 | - | 1 | 865 | ||
| BC022137.1 | - | 1 | 796 | |||
| BC030734.1 | - | 1 | 669 | |||
| BC034832.1 | - | 1 | 828 | |||