siRNA Details
| Gene Symbol: Limd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013860.2 | NP_038888.2 | 1 | 1749 | ||
| GenBank mRNA | AJ132409.1 | - | 1 | 1738 | ||
| AK004806.1 | - | 1 | 1053 | |||
| AK047198.1 | - | 1 | 1743 | |||
| AK083970.1 | - | 1 | 1710 | |||
| AK191283.1 | - | 1 | 65 | |||
| AK191584.1 | - | 1 | 65 | |||
| AK192393.1 | - | 1 | 65 | |||
| AK195125.1 | - | 1 | 73 | |||
| AK198802.1 | - | 1 | 65 | |||
| AK200245.1 | - | 1 | 65 | |||
| AK203953.1 | - | 1 | 73 | |||
| AK208965.1 | - | 1 | 65 | |||
| BC056449.1 | - | 1 | 1624 | 5703026 | ||

Limd1
Genomic Map