siRNA Details
| Gene Symbol: Limd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013860.2 | NP_038888.2 | 1 | 1677 | ||
| GenBank mRNA | AJ132409.1 | - | 1 | 1666 | ||
| AK004806.1 | - | 1 | 981 | |||
| AK047198.1 | - | 1 | 1671 | |||
| AK083970.1 | - | 1 | 1638 | |||
| BC056449.1 | - | 1 | 1552 | 5703026 | ||

Limd1
Genomic Map