siRNA Details
| Gene Symbol: Gtse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168672.1 | NP_001162143.1 | 9 | 1730 | ||
| NM_013882.2 | NP_038910.1 | 8 | 1700 | |||
| GenBank mRNA | AJ222580.1 | - | 8 | 1670 | ||
| AK081582.1 | - | 9 | 1693 | |||
| BC027213.1 | - | 8 | 1701 | 4039841 | ||

Gtse1
Genomic Map