siRNA Details
| Gene Symbol: Gtse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168672.1 | NP_001162143.1 | 4 | 424 | ||
| NM_013882.2 | NP_038910.1 | 3 | 394 | |||
| GenBank mRNA | AJ222580.1 | - | 3 | 364 | ||
| AK012870.1 | - | 4 | 425 | |||
| AK081582.1 | - | 4 | 387 | |||
| BC027213.1 | - | 3 | 395 | 4039841 | ||

Gtse1
Genomic Map