siRNA Details
| Gene Symbol: Dnajc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253685.1 | NP_001240614.1 | 4 | 316 | ||
| NM_013888.3 | NP_038916.1 | 4 | 453 | |||
| GenBank mRNA | AF132906.2 | - | 4 | 402 | ||
| AK002649.1 | - | 2 | 141 | |||
| AK032207.1 | - | 3 | 1822 | |||
| BC013501.1 | - | 4 | 360 | 4222828 | ||
| BC071211.1 | - | 3 | 1718 | |||
| BF453706.1 | - | 4 | 316 | |||

Dnajc12
Genomic Map