siRNA Details
| Gene Symbol: Adamts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013906.2 | NP_038934.2 | 8, 9 | 2424 | ||
| GenBank mRNA | AF175282.1 | - | 8, 9 | 2427 | ||
| AK043875.1 | - | 1396 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts8| Gene Symbol: Adamts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013906.2 | NP_038934.2 | 8, 9 | 2424 | ||
| GenBank mRNA | AF175282.1 | - | 8, 9 | 2427 | ||
| AK043875.1 | - | 1396 | ||||