siRNA Details
| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013910.2 | NP_038938.1 | 5 | 679 | ||
| GenBank mRNA | AK129479.1 | - | 16 | 2116 | ||
| BC057622.1 | - | 5 | 674 | 6402440 | ||
| BC065090.1 | - | 16 | 2315 | 6851421 | ||
| BC068239.1 | - | 13 | 1869 | |||

Kdm2b
Genomic Map