siRNA Details
| Gene Symbol: Tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133673.3 | NP_598434.2 | 2 | 463 | ||
| GenBank mRNA | AJ297743.1 | - | 2 | 493 | ||
| AK017290.1 | - | 2 | 482 | |||
| AK028859.1 | - | 2 | 277 | |||
| AK054404.1 | - | 2 | 494 | |||
| AK149754.1 | - | 2 | 484 | |||
| AK159843.1 | - | 2 | 462 | |||
| AK169692.1 | - | 2 | 486 | |||
| BC018456.1 | - | 2 | 462 | 3582246 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001039197, FQ222570, BC111704.

Tor1b
Genomic Map