siRNA Details
| Gene Symbol: Slc46a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021053.4 | NP_066395.3 | 1 | 293 | ||
| GenBank mRNA | AF148145.1 | - | 1 | 265 | ||
| AK014784.1 | - | 1 | 280 | |||
| AK017363.1 | - | 1 | 293 | |||
| AK040063.1 | - | 294 | ||||
| BC140962.1 | - | 1 | 226 | 9055574 | ||

Slc46a2
Genomic Map