siRNA Details
| Gene Symbol: Usp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013918.2 | NP_038946.2 | 13 | 1374 | ||
| GenBank mRNA | AF170563.1 | - | 13 | 1520 | ||
| BC048171.1 | - | 13 | 1374 | 6822095 | ||
| BC063059.1 | - | 13 | 1374 | 6822095 | ||

Usp25
Genomic Map