siRNA Details
| Gene Symbol: Zfp354c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013922.4 | NP_038950.3 | 5 | 716 | ||
| GenBank mRNA | AF192804.1 | - | 4 | 511 | ||
| AK086760.1 | - | 5 | 679 | |||
| AK145882.1 | - | 5 | 710 | |||
| AK220194.1 | - | 5 | 684 | |||
| BC079908.1 | - | 5 | 716 | 30654724 | ||

Zfp354c
Genomic Map