siRNA Details
| Gene Symbol: Schip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113420.1 | NP_001106891.1 | 5 | 728 | ||
| NM_001113421.1 | NP_001106892.1 | 6 | 1412 | |||
| NM_001282044.1 | NP_001268973.1 | 6 | 1395 | |||
| NM_001282045.1 | NP_001268974.1 | 6 | 1253 | |||
| NM_013928.5 | NP_038956.2 | 5 | 818 | |||
| GenBank mRNA | AF145716.1 | - | 5 | 924 | ||
| AK021361.1 | - | 5 | 679 | |||
| AK083464.1 | - | 6 | 1238 | |||
| AK146930.1 | - | 6 | 1369 | |||
| AK168005.1 | - | 6 | 1322 | |||
| BC060529.1 | - | 5 | 752 | |||
| EU163407.1 | - | 6 | 1412 | |||
| EU163408.1 | - | 5 | 728 | |||
| KM233715.1 | - | 5 | 674 | |||
| KM233716.1 | - | 5 | 679 | |||
| Gene Symbol: Gm21949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113419.2 | NP_001106890.1 | 9 | 1590 | ||
| GenBank mRNA | KJ941154.1 | - | 8 | 894 | ||

Schip1
Genomic Map