siRNA Details
| Gene Symbol: Pik3cg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146200.1 | NP_001139672.1 | 4 | 2356 | ||
| NM_001146201.1 | NP_001139673.1 | 4 | 2412 | |||
| NM_020272.2 | NP_064668.2 | 4 | 2533 | |||
| GenBank mRNA | AF208345.1 | - | 3 | 2212 | ||
| AJ249280.1 | - | 4 | 2533 | |||
| AK040543.1 | - | 4 | 2318 | |||
| AK132553.1 | - | 4 | 2358 | |||
| AK137470.1 | - | 4 | 2265 | |||
| AK137620.1 | - | 4 | 2414 | |||
| AK143238.1 | - | 1 | 37 | |||
| BC051246.1 | - | 4 | 2273 | 6514979 | ||

Pik3cg
Genomic Map