siRNA Details
| Gene Symbol: Aass | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013930.4 | NP_038958.2 | 13, 14 | 1528 | ||
| GenBank mRNA | AJ224761.1 | - | 12, 13 | 1419 | ||
| AK044323.1 | - | 13, 14 | 1509 | |||
| AK098141.1 | - | 13, 14 | 1469 | |||
| AK136216.1 | - | 13, 14 | 1477 | |||
| AK149400.1 | - | 13, 14 | 1453 | |||
| BC005420.1 | - | 13, 14 | 1514 | 3600654 | ||

Aass
Genomic Map