siRNA Details
| Gene Symbol: Gimap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174960.2 | NP_777620.1 | 2 | 181 | ||
| GenBank mRNA | AK041336.1 | - | 2 | 170 | ||
| BB618178.1 | - | 2 | 181 | |||
| BC038043.1 | - | 2 | 103 | |||
| BC096680.1 | - | 2 | 165 | 1345477 | ||

Gimap9
Genomic Map