siRNA Details
| Gene Symbol: Gpr82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175669.3 | NP_783600.1 | 4 | 291 | ||
| GenBank mRNA | AK033761.1 | - | 4 | 293 | ||
| BC116704.1 | - | 4 | 286 | 40104936 | ||
More Information
Set Membership: |
|

Gpr82
Genomic Map