siRNA Details
| Gene Symbol: Fchsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175684.4 | NP_783615.2 | 8 | 629 | ||
| GenBank mRNA | AK079461.1 | - | 2 | 101 | ||
| AK154732.1 | - | 8 | 663 | |||
| BC052886.1 | - | 8 | 660 | |||
| BC057367.1 | - | 8 | 629 | 6827858 | ||

Fchsd1
Genomic Map