siRNA Details
| Gene Symbol: Fchsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175684.4 | NP_783615.2 | 17 | 1684 | ||
| GenBank mRNA | AK079461.1 | - | 11 | 1156 | ||
| AK154732.1 | - | 17 | 1719 | |||
| BC052886.1 | - | 17 | 1715 | |||
| BC057367.1 | - | 17 | 1684 | 6827858 | ||

Fchsd1
Genomic Map