siRNA Details
Gene Symbol: A230046K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033375.2 | NP_001028547.2 | 17 | 1707 | ||
GenBank mRNA | AK038582.1 | - | 17 | 1665 | ||
AK043911.1 | - | 17 | 1678 | |||
AK129269.1 | - | 12 | 1237 | |||
AK145000.1 | - | 17 | 1689 | |||
AK166233.1 | - | 17 | 1680 | |||
BC020314.2 | - | 14 | 1342 | |||
BC026467.1 | - | 6 | 668 | |||
BC145215.1 | - | 17 | 1902 |
