siRNA Details
Gene Symbol: A230046K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033375.2 | NP_001028547.2 | 31, 32 | 3446 | ||
GenBank mRNA | AK038582.1 | - | 31, 32 | 3404 | ||
AK043911.1 | - | 31, 32 | 3417 | |||
AK129269.1 | - | 26, 27 | 2976 | |||
AK145000.1 | - | 31, 32 | 3428 | |||
AK161247.1 | - | 8, 9 | 894 | |||
AK166233.1 | - | 31, 32 | 3419 | |||
BC020314.2 | - | 28, 29 | 3081 | |||
BC026467.1 | - | 20, 21 | 2407 | |||
BC145215.1 | - | 31, 32 | 3641 |
