siRNA Details
Gene Symbol: Snhg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175692.3 | NP_783623.2 | 2, 3 | 496 | ||
GenBank mRNA | AK080745.1 | - | 2, 3 | 454 | ||
BC030414.1 | - | 2, 3 | 412 | 5366924 | ||
BC049241.1 | - | 410 | 4505955 |
