siRNA Details
| Gene Symbol: Atg2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029654.4 | NP_083930.5 | 6 | 1261 | ||
| GenBank mRNA | AK032840.1 | - | 4 | 527 | ||
| AK083213.1 | - | 6 | 1221 | |||
| AK085790.1 | - | 6 | 1208 | |||
| AK086802.1 | - | 6 | 1262 | |||
| BC062182.1 | - | 6 | 1231 | 6505402 | ||

Atg2b
Genomic Map