siRNA Details
| Gene Symbol: Fndc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207636.2 | NP_997519.2 | 5 | 788 | ||
| GenBank mRNA | AK007062.1 | - | 1 | 29 | ||
| AK032733.1 | - | 5 | 760 | |||
| AK048586.1 | - | 5 | 791 | |||
| AK089380.1 | - | 5 | 752 | |||
| AK154790.1 | - | 5 | 558 | |||
| AK172458.1 | - | 5 | 652 | |||
| BC144880.1 | - | 5 | 596 | |||
| DQ192036.1 | - | 5 | 789 | |||

Fndc3a
Genomic Map