siRNA Details
| Gene Symbol: Fndc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207636.2 | NP_997519.2 | 9 | 1314 | ||
| GenBank mRNA | AK032733.1 | - | 9 | 1286 | ||
| AK048586.1 | - | 9 | 1317 | |||
| AK089380.1 | - | 9 | 1279 | |||
| AK154790.1 | - | 9 | 1083 | |||
| AK172458.1 | - | 9 | 1178 | |||
| BC144880.1 | - | 9 | 1122 | |||
| DQ192036.1 | - | 9 | 1315 | |||

Fndc3a
Genomic Map