siRNA Details
| Gene Symbol: Lrtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176920.4 | NP_795894.1 | 2 | 469 | ||
| GenBank mRNA | AK044492.1 | - | 2 | 423 | ||
| AK138618.1 | - | 2 | 287 | |||
| BC099587.1 | - | 2 | 267 | 30289234 | ||
| BC132594.1 | - | 2 | 173 | 40130871 | ||
| BC145196.1 | - | 2 | 173 | |||
| BC145990.1 | - | 2 | 173 | 40131293 | ||

Lrtm1
Genomic Map