siRNA Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198308.1 | NP_938050.1 | 3 | 386 | ||
| GenBank mRNA | AK029575.1 | - | 3 | 477 | ||
| AK129472.1 | - | 3 | 490 | |||
| AY223867.1 | - | 3 | 386 | |||
| BC120745.1 | - | 2 | 316 | 40129668 | ||
| BC125425.1 | - | 2 | 316 | 40129940 | ||

Pdpr
Genomic Map