siRNA Details
| Gene Symbol: Zfp750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178763.4 | NP_848878.1 | 2 | 1433 | ||
| GenBank mRNA | AK028879.1 | - | 2 | 1382 | ||
| AK030690.1 | - | 2 | 1388 | |||
| AK037185.1 | - | 2 | 1394 | |||
| AK037240.1 | - | 2 | 1379 | |||
| AK076268.1 | - | 2 | 1427 | |||
| AK140792.1 | - | 1426 | ||||
| BC080834.1 | - | 2 | 1404 | |||
| BC150905.1 | - | 1 | 1287 | 9087816 | ||

Zfp750
Genomic Map