siRNA Details
| Gene Symbol: Lig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176953.3 | NP_795927.2 | 2 | 1660 | ||
| GenBank mRNA | AK030029.1 | - | 1542 | |||
| AK083353.1 | - | 2 | 1540 | |||
| AK148081.1 | - | 1636 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lig4| Gene Symbol: Lig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176953.3 | NP_795927.2 | 2 | 1660 | ||
| GenBank mRNA | AK030029.1 | - | 1542 | |||
| AK083353.1 | - | 2 | 1540 | |||
| AK148081.1 | - | 1636 | ||||