siRNA Details
| Gene Symbol: Nt5dc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176968.4 | NP_795942.2 | 12 | 1365 | ||
| GenBank mRNA | AK049656.1 | - | 11 | 1280 | ||
| AK077876.1 | - | 12 | 1366 | |||
| AK145666.1 | - | 12 | 1366 | |||
| AK164144.1 | - | 12 | 1306 | |||
| AK171761.1 | - | 12 | 1375 | |||
| BC098476.1 | - | 12 | 1325 | 30051251 | ||
| BC125276.1 | - | 12 | 1364 | 40107511 | ||

Nt5dc1
Genomic Map