siRNA Details
Gene Symbol: Podxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176973.4 | NP_795947.3 | 5 | 1265 | ||
GenBank mRNA | AK051761.1 | - | 5 | 1468 | ||
BC033384.1 | - | 5 | 1265 | 5362581 |
Gene Symbol: Gm15612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133241.1 | - | 3 | 1448 |
