siRNA Details
Gene Symbol: Podxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176973.4 | NP_795947.3 | 7 | 1548 | ||
GenBank mRNA | AK051761.1 | - | 7 | 1751 | ||
BC033384.1 | - | 7 | 1548 | 5362581 |
Gene Symbol: Gm15612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133241.1 | - | 2 | 300 |
