siRNA Details
| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178767.5 | NP_848882.2 | 2 | 612 | ||
| GenBank mRNA | AK039191.1 | - | 2 | 183 | ||
| AK040702.1 | - | 2 | 613 | |||
| AK050246.1 | - | 2 | 473 | |||
| AK163155.1 | - | 2 | 393 | |||
| BC038044.1 | - | 2 | 512 | |||

Agmo
Genomic Map