siRNA Details
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028127.3 | NP_082403.2 | 7 | 812 | ||
| GenBank mRNA | AK029709.1 | - | 6 | 571 | ||
| AK054457.1 | - | 7 | 1173 | |||
| BC053929.1 | - | 7 | 812 | 6389695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmd6| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028127.3 | NP_082403.2 | 7 | 812 | ||
| GenBank mRNA | AK029709.1 | - | 6 | 571 | ||
| AK054457.1 | - | 7 | 1173 | |||
| BC053929.1 | - | 7 | 812 | 6389695 | ||