siRNA Details
| Gene Symbol: Simc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176987.4 | NP_795961.3 | 9 | 3855 | ||
| GenBank mRNA | AK028926.1 | - | 9 | 1520 | ||
| AK052561.1 | - | 8 | 1649 | |||
| AK166176.1 | - | 8 | 1459 | |||
| BC023054.1 | - | 4 | 475 | |||
| BC059035.1 | - | 8 | 2114 | |||
| BC094550.1 | - | 8 | 1666 | |||

Simc1
Genomic Map