siRNA Details
Gene Symbol: Simc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176987.4 | NP_795961.3 | 7, 8 | 3654 | ||
GenBank mRNA | AK028926.1 | - | 7, 8 | 1319 | ||
AK052561.1 | - | 6, 7 | 1448 | |||
AK166176.1 | - | 6, 7 | 1258 | |||
BC023054.1 | - | 2, 3 | 274 | |||
BC059035.1 | - | 6, 7 | 1913 | |||
BC094550.1 | - | 6, 7 | 1465 |
