siRNA Details
| Gene Symbol: Simc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176987.4 | NP_795961.3 | 10 | 3903 | ||
| GenBank mRNA | AK028926.1 | - | 10 | 1568 | ||
| AK052561.1 | - | 9 | 1697 | |||
| AK166176.1 | - | 9 | 1507 | |||
| BC023054.1 | - | 5 | 523 | |||
| BC059035.1 | - | 9 | 2162 | |||
| BC094550.1 | - | 9 | 1714 | |||

Simc1
Genomic Map