siRNA Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030180.2 | NP_084456.2 | 9 | 1602 | ||
| GenBank mRNA | AK036864.1 | - | 8 | 991 | ||
| AK047620.1 | - | 9 | 1602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp54| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030180.2 | NP_084456.2 | 9 | 1602 | ||
| GenBank mRNA | AK036864.1 | - | 8 | 991 | ||
| AK047620.1 | - | 9 | 1602 | |||